missing translation for 'onlineSavingsMsg'
Learn More

Arbutin, 98%

Catalog No. p-7077569
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455640050 5 g
AC455640250 25 g
AC455641000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC455640050 Supplier Thermo Scientific Chemicals Supplier No. 455640050
Only null left

CAS: 497-76-7 | C12H16O7 | 272.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 497-76-7
Molecular Formula C12H16O7
Molecular Weight (g/mol) 272.25
MDL Number MFCD00016915
InChI Key BJRNKVDFDLYUGJ-UHFFFAOYNA-N
Synonym arbutin, uvasol, 4-hydroxyphenyl beta-d-glucopyranoside, ursin, beta-arbutin, arbutoside, arbutine, arbutyne, ursi, p-arbutin
PubChem CID 440936
ChEBI CHEBI:18305
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-hydroxyphenoxy)oxane-3,4,5-triol
SMILES OCC1OC(OC2=CC=C(O)C=C2)C(O)C(O)C1O

Specifications

Melting Point 195.0°C to 201.0°C
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: 50g/L. Other solubilities: soluble in alcohols
Formula Weight 272.25
Percent Purity 98%
Chemical Name or Material Arbutin
EINECSNumber 207-850-3

RUO – Research Use Only