missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Aramid Fiber/Nomex™ High Heat Gloves

Catalog No. 19148794
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19148794 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19148794 Supplier Wells Lamont™ Supplier No. 2610L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Seamless knit, cotton-lined Aramid Fiber/Nomex™ High Heat Gloves are capable of withstanding extreme temperatures up to 500°F.

  • High heat gloves are capable of withstanding extreme temperatures up to 500°F
  • Cotton lining provides comfort and additional heat resistance
  • Gloves are laundered and retain heat resistance even after repeated washings
  • Made in the USA

Specifications

Certifications/Compliance ANSI Flame Rating Level 4
Construction Seamless Knit
Coating Material Uncoated
Color White, Yellow
Cut Resistance Rating ANSI A3
Hand Ambidextrous
Material Aramid Fiber, Nomex
Product Type High Heat Gloves
Size Large
Temperature (English) Max. 500°F
Temperature (Metric) Max 260°C
For Use With (Application) Food Processing, Foundry, Sheet Metal, and Steel Manufacturing
Glove Cuff Style Continuous Knit
Hem Color White
Liner Material Cotton
Series 2610
Show More Show Less