missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arachidonic acid, >99%, MP Biomedicals™
$75.15
Chemical Identifiers
| CAS | 506-32-1 |
|---|---|
| Molecular Formula | C20H32O2 |
| Molecular Weight (g/mol) | 304.474 |
| InChI Key | YZXBAPSDXZZRGB-DOFZRALJSA-N |
| Synonym | arachidonic acid, arachidonate, immunocytophyte, all-z-5,8,11,14-eicosatetraenoic acid, 5z,8z,11z,14z-icosa-5,8,11,14-tetraenoic acid, cis-5,8,11,14-eicosatetraenoic acid, 5,8,11,14-eicosatetraenoic acid, all-z, all-cis-5,8,11,14-eicosatetraenoic acid, unii-27yg812j1i, 5z,8z,11z,14z-eicosatetraenoic acid |
| PubChem CID | 444899 |
| ChEBI | CHEBI:15843 |
| IUPAC Name | (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoic acid |
| SMILES | CCCCCC=CCC=CCC=CCC=CCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN19462510
|
N/A | 10 mg |
Each for $75.15
|
|
|||||
Chemical Identifiers
| 506-32-1 | |
| 304.474 | |
| arachidonic acid, arachidonate, immunocytophyte, all-z-5,8,11,14-eicosatetraenoic acid, 5z,8z,11z,14z-icosa-5,8,11,14-tetraenoic acid, cis-5,8,11,14-eicosatetraenoic acid, 5,8,11,14-eicosatetraenoic acid, all-z, all-cis-5,8,11,14-eicosatetraenoic acid, unii-27yg812j1i, 5z,8z,11z,14z-eicosatetraenoic acid | |
| CHEBI:15843 | |
| CCCCCC=CCC=CCC=CCC=CCCCC(=O)O |
| C20H32O2 | |
| YZXBAPSDXZZRGB-DOFZRALJSA-N | |
| 444899 | |
| (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoic acid |
Specifications
| 506-32-1 | |
| 0.922g/mL | |
| arachidonic acid, arachidonate, immunocytophyte, all-z-5,8,11,14-eicosatetraenoic acid, 5z,8z,11z,14z-icosa-5,8,11,14-tetraenoic acid, cis-5,8,11,14-eicosatetraenoic acid, 5,8,11,14-eicosatetraenoic acid, all-z, all-cis-5,8,11,14-eicosatetraenoic acid, unii-27yg812j1i, 5z,8z,11z,14z-eicosatetraenoic acid | |
| CCCCCC=CCC=CCC=CCC=CCCCC(=O)O | |
| 304.474 | |
| CHEBI:15843 | |
| >99% | |
| Arachidonic acid |
| Colorless | |
| C20H32O2 | |
| YZXBAPSDXZZRGB-DOFZRALJSA-N | |
| (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoic acid | |
| 444899 | |
| 304.47 | |
| Liquid |