missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arachidic Acid 98.0+%, TCI America™
$32.28 - $235.43
Chemical Identifiers
| CAS | 506-30-9 |
|---|---|
| Molecular Formula | C20H40O2 |
| Molecular Weight (g/mol) | 312.54 |
| MDL Number | MFCD00002755 |
| InChI Key | VKOBVWXKNCXXDE-UHFFFAOYSA-N |
| Synonym | arachidic acid, eicosanoic acid, arachic acid, n-eicosanoic acid, arachidinic acid, arachidate, eicosanoate, unii-pqb8mjd4rb, pqb8mjd4rb, n-eicosanoate |
| PubChem CID | 10467 |
| ChEBI | CHEBI:28822 |
| IUPAC Name | icosanoic acid |
| SMILES | CCCCCCCCCCCCCCCCCCCC(O)=O |
Chemical Identifiers
| 506-30-9 | |
| 312.54 | |
| VKOBVWXKNCXXDE-UHFFFAOYSA-N | |
| 10467 | |
| icosanoic acid |
| C20H40O2 | |
| MFCD00002755 | |
| arachidic acid, eicosanoic acid, arachic acid, n-eicosanoic acid, arachidinic acid, arachidate, eicosanoate, unii-pqb8mjd4rb, pqb8mjd4rb, n-eicosanoate | |
| CHEBI:28822 | |
| CCCCCCCCCCCCCCCCCCCC(O)=O |
Specifications
| 506-30-9 | |
| White | |
| C20H40O2 | |
| 1 g | |
| VKOBVWXKNCXXDE-UHFFFAOYSA-N | |
| icosanoic acid | |
| 10467 | |
| 312.54 | |
| Crystalline Powder |
| 76°C | |
| 303°C | |
| MFCD00002755 | |
| arachidic acid, eicosanoic acid, arachic acid, n-eicosanoic acid, arachidinic acid, arachidate, eicosanoate, unii-pqb8mjd4rb, pqb8mjd4rb, n-eicosanoate | |
| CCCCCCCCCCCCCCCCCCCC(O)=O | |
| 312.54 | |
| CHEBI:28822 | |
| ≥98.0% (GC,T) | |
| Arachidic Acid |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : JX3780000
TSCA : Yes