missing translation for 'onlineSavingsMsg'
Learn More

Arachidic Acid 98.0+%, TCI America™

Catalog No. E000625G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E000625G 25 g
E00061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E000625G Supplier TCI America Supplier No. E000625G

Chemical Identifiers

CAS 506-30-9
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.54
MDL Number MFCD00002755
InChI Key VKOBVWXKNCXXDE-UHFFFAOYSA-N
Synonym arachidic acid, eicosanoic acid, arachic acid, n-eicosanoic acid, arachidinic acid, arachidate, eicosanoate, unii-pqb8mjd4rb, pqb8mjd4rb, n-eicosanoate
PubChem CID 10467
ChEBI CHEBI:28822
IUPAC Name icosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 76°C
Color White
Boiling Point 303°C
Quantity 25 g
Formula Weight 312.54
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Arachidic Acid
EINECSNumber (2)-0608
RTECSNumber JX3780000
TSCA Yes