missing translation for 'onlineSavingsMsg'
Learn More

Arachidic acid, 98%

Catalog No. AAA1961603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1961603 1 g
AAA1961606 5 g
AAA1961614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1961603 Supplier Thermo Scientific Chemicals Supplier No. A1961603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 506-30-9 | C20H40O2 | 312.54 g/mol

Arachidic acid is used in the manufacture of pharmaceuticals, soaps, cosmetics, and food packaging because of its surfactant-like properties. Arachidic acid is a saturated fatty acid with a 20 carbon chain. Diets rich in saturated fats like arachidic acid are associated with increased levels of serum low density lipoproteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Arachidic acid is used in the manufacture of pharmaceuticals, soaps, cosmetics, and food packaging because of its surfactant-like properties. Arachidic acid is a saturated fatty acid with a 20 carbon chain. Diets rich in saturated fats like arachidic acid are associated with increased levels of serum low density lipoproteins.

Solubility
Soluble in chloroform, ether, ethanol, and DMF. Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 506-30-9
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.54
MDL Number MFCD00002755
InChI Key VKOBVWXKNCXXDE-UHFFFAOYSA-N
Synonym arachidic acid, eicosanoic acid, arachic acid, n-eicosanoic acid, arachidinic acid, arachidate, eicosanoate, unii-pqb8mjd4rb, pqb8mjd4rb, n-eicosanoate
PubChem CID 10467
ChEBI CHEBI:28822
IUPAC Name icosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 74°C to 76°C
Quantity 1 g
Merck Index 14,764
Solubility Information Soluble in chloroform,ether,ethanol,and DMF. Insoluble in water.
Formula Weight 312.54
Percent Purity 98%
Chemical Name or Material Arachidic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 208-031-3
RTECSNumber JX3780000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only