Learn More
Electron Microscopy Glutaraldehyde 8% Aqueous Solution, 111-30-8, MFCD00007025, 10x2 mL DFS Item
Supplier:  Electron Microscopy Sciences 16019
Formula: OCHCH2CH2CH2CHO, MW: 100.12. After glutaraldehyde has been purified to EM grade, it has a high risk of polymerizing if it is not handled properly. Electron Microscopy Sciences takes special precautions to assure a polymer-free, pure glutaraldehyde. 4 to (-18)C.
Catalog No. 5026210
Only null left
Explore available promotions