missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Aquasearcher™ pH Meter AB41PH
Description
- Equipped with a large 6.5 in. LCD display and a touch keypad
- Features a 1000 item memory and password management for GLP documentation
- Provides connectivity and data output capabilities with RS232 and USB interface
- i-Steward ensures repeatable and consistent measurement accuracy
- Intuitive user interface makes changing the parameters, executing setup, and performing calibration as simple as using a smartphone
- Choose between just the AB41PH meter with electrode holder and a full kit including AB41PH meter, ST410 electrode, STTEMP probe, electrode holder, AS20 magnetic stirrer, and 4 pH buffer mini kits (50 mL)
Specifications
Specifications
| Data Management Details | Datalog with Time and Date Stamp |
| Accuracy (pH) | ±0.002 |
| Accuracy (Temperature) | ±0.3°C |
| Accuracy (mV) | ±0.3 |
| Outputs | USB, RS-232 (PC), RS-232 (Printer) |
| Includes | AB41PH Meter, Electrode Holder |
| Measurement Modes | Auto-stop, Timed, Continuous |
| Calibration | Offset |
| Range (pH) | -2 to 20 |
| Resolution (pH) | 0.001, 0.01, 0.1 |
| Show More |