missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Aquasearcher™ pH Meter AB41PH

Catalog No. 01922367
Change view
Click to view available options
Type:
Meter with pH Kit
pH Meter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01922367 pH Meter
01922366 Meter with pH Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01922367 Supplier OHAUS™ Supplier No. 30589830

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Aquasearcher™ pH Meter AB41PH is an advanced, research-grade benchtop pH meter with electrode, offering accurate, repeatable results with selectable resolution and an intelligent i-Steward system.

  • Equipped with a large 6.5 in. LCD display and a touch keypad
  • Features a 1000 item memory and password management for GLP documentation
  • Provides connectivity and data output capabilities with RS232 and USB interface
  • i-Steward ensures repeatable and consistent measurement accuracy
  • Intuitive user interface makes changing the parameters, executing setup, and performing calibration as simple as using a smartphone
  • Choose between just the AB41PH meter with electrode holder and a full kit including AB41PH meter, ST410 electrode, STTEMP probe, electrode holder, AS20 magnetic stirrer, and 4 pH buffer mini kits (50 mL)

Specifications

Data Management Details Datalog with Time and Date Stamp
Accuracy (pH) ±0.002
Accuracy (Temperature) ±0.3°C
Accuracy (mV) ±0.3
Outputs USB, RS-232 (PC), RS-232 (Printer)
Includes AB41PH Meter, Electrode Holder
Measurement Modes Auto-stop, Timed, Continuous
Calibration Offset
Range (pH) -2 to 20
Resolution (pH) 0.001, 0.01, 0.1
Range (mV) ±2000.0
Calibration Points Up to 5
Temperature Range (Metric) -10°C to +125°C
Display Type LCD, 6.5 in. Segment and Dot Matrix
Backlight Option LCD
Data Points 1000 Sets
Log Function Type 1000 Measurement, 10 Calibration
Type pH Meter
Show More Show Less