missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Aquasearcher™ pH Meter AB33PH
The Aquasearcher™ pH Meter AB33PH is a highly reliable and user-friendly pH benchtop meter with electrode (ST310) for standard laboratory applications. Designed to measure pH and oxidation-reduction potential with temperature measurements.
$1,560.02 - $1,772.74
Specifications
| Accuracy (mV) | ±1 |
|---|---|
| Accuracy (pH) | ±0.01 |
| Accuracy (Temperature) | ±0.5°C |
| Data Management Details | Datalog with Time and Date Stamp |
| Outputs | USB, RS-232 (PC), RS-232 (Printer) |
Description
- Equipped with a large 6.5 in. LCD display and a touch keypad
- Features a 1000 item memory for pH measurements and calibration trail
- Provides connectivity and data output capabilities with RS232 and USB interface
- i-Steward monitors the condition of electrodes to ensure repeatable and consistent measurement accuracy
- Intuitive user interface makes changing the parameters, executing setup, and performing calibration as simple as using a smartphone
- Auto endpoint mode and auto buffer recognition makes calibration easy
- Choose between just the AB33PH meter with electrode holder and a full kit including AB33PH meter, ST310 electrode, electrode holder, 4 pH buffer mini kits (50 mL)
Specifications
| ±1 | |
| ±0.5°C | |
| USB, RS-232 (PC), RS-232 (Printer) | |
| Linear | |
| 0.01, 0.1 | |
| Up to 3 | |
| LCD, 6.5 in. Segment and Dot Matrix | |
| 1000 Sets |
| ±0.01 | |
| Datalog with Time and Date Stamp | |
| Auto-stop, Continuous | |
| -2 to 16 | |
| ±2000.0 | |
| -5°C to +110°C | |
| LCD | |
| 1000 Measurement, Last Calibration |