missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AquaMet 95.0+%, TCI America™
$516.35 - $1549.03
Chemical Identifiers
| CAS | 1414707-08-6 |
|---|---|
| Molecular Formula | C39H55Cl3N4ORu |
| MDL Number | MFCD28411647 |
| Synonym | [1,3-Bis(2,4,6-trimethylphenyl)-4-[(4-ethyl-4-methylpiperazinium-1-yl)methyl]-2-imidazolidinylidene]dichloro(2-isopropoxybenzylidene)ruthenium(VI) Chloride |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A3009100MG
|
TCI America
A3009100MG |
100 mg |
Each for $516.35
|
|
|||||
|
A3009500MG
|
TCI America
A3009500MG |
500 mg |
Each for $1,549.03
|
|
|||||
Chemical Identifiers
| 1414707-08-6 | |
| C39H55Cl3N4ORu | |
| MFCD28411647 | |
| [1,3-Bis(2,4,6-trimethylphenyl)-4-[(4-ethyl-4-methylpiperazinium-1-yl)methyl]-2-imidazolidinylidene]dichloro(2-isopropoxybenzylidene)ruthenium(VI) Chloride |
Specifications
| 1414707-08-6 | |
| C39H55Cl3N4ORu | |
| 100 mg | |
| 803.32 | |
| Crystalline Powder |
| Green-Yellow | |
| MFCD28411647 | |
| [1,3-Bis(2,4,6-trimethylphenyl)-4-[(4-ethyl-4-methylpiperazinium-1-yl)methyl]-2-imidazolidinylidene]dichloro(2-isopropoxybenzylidene)ruthenium(VI) Chloride | |
| ≥95.0% (HPLC) | |
| AquaMet |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator