Learn More
Karl Fischer Aqualine™ Sodium Tartrate-2H2O, for Karl Fischer Titration, Fisher Chemical
Solid standard
Supplier:  Fisher Chemical K277048
Catalog No. K2770-100
Only null left
Explore available promotions
Description
Description
  • Primary solid water standard for Karl Fischer volumetric analysis
  • Contains 15.66% ± 0.05% water
Specifications
Chemical Identifiers
0.0
Product Suggestions
Product Suggestions