Learn More
Decagon Devices AquaLab TDL 2, Tunable Diode Laser Sensor, 0.0050 aw Accuracy, 5 min Read Time
Catalog No. NC2066601
Encompass
The AquaLab TDL 2 is an advanced water activity meter that ensures precise measurement of water activity in various complex samples. It utilizes tunable diode laser technology for fast readings, high accuracy of 0.0050 aw, and low maintenance through a sealed design. Ideal for industries requiring quality assurance, it complies with international standards, making it a reliable choice for professionals needing accurate moisture control.
Catalog No. NC2066601 Supplier Decagon Devices Supplier No. 40393
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The AquaLab TDL 2 is an advanced water activity meter that ensures precise measurement of water activity in various complex samples. It utilizes tunable diode laser technology for fast readings, high accuracy of 0.0050 aw, and low maintenance through a sealed design. Ideal for industries requiring quality assurance, it complies with international standards, making it a reliable choice for professionals needing accurate moisture control.