missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ APTS (8-Aminopyrene-1,3,6-Trisulfonic Acid, Trisodium Salt)

Catalog No. A6257
Encompass
Change view
Click to view available options
Quantity:
10 mg
Packaging:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A6257 10 mg 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A6257 Supplier Molecular Probes™ Supplier No. A6257
Only null left

APTS (8-Aminopyrene-1,3,6-Trisulfonic Acid, Trisodium Salt)

The aromatic amine of APTS (8-aminopyrene-1,3,6-trisulfonic acid, trisodium salt) can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3) to form new biotinylated probes.

Specifications

Melting Point >250°C
Color Yellow
Packaging 1 Each
Quantity 10 mg
Physical Form Solid

For Research Use Only. Not for use in diagnostic procedures.