Learn More
Fisherbrand™ Drosophila Products Jazz-Mix™ Drosophila Food
Catalog No. AS153
Change view
Click to view available options
Type:
Jazzmix Drosophila Food
Catalog No. Type
AS153 Jazzmix Drosophila Food
1 options
Catalog No. AS153 Supplier Fisherbrand™ Supplier No. JAZZMIX10LB
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Simply add water—conveniently packaged as a complete and quick-to-prepare mixture.
  • Consists of brown sugar, corn meal, yeast, agar, benzoic acid, methyl paraben and propionic acid
Notes: Product is not made for human consumption. Yield may vary based on evaporation while boiling.
Order Info
Packaged in a heavy twist-tied plastic bag inside a durable corrugated box
Specifications
Type Jazzmix Drosophila Food
Weight (Metric) 4.5 kg
Weight (English) 10 lb.