missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ RapidLINK™ Software v1.0 Upgrade

Catalog No. A41820
Change view
Click to view available options
Quantity:
1 upgrade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A41820 1 upgrade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A41820 Supplier Applied Biosystems™ Supplier No. A41820

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Applied Biosystems RapidLINK Software v1.0 is part of the Applied Biosystems RapidHIT ID System.

NOTE: For use with the Applied Biosystems-branded RapidHIT ID System only (Cat. No. A41810).

Applied Biosystems RapidLINK Software v1.0 is part of the Applied Biosystems RapidHIT ID System. This fast and simple system produces trusted lab-quality forensic DNA profiles in 90 minutes that are aggregated within the software for direct upload to the database of choice or for further review and analysis. This data management software expedites identity confirmation securely and efficiently.

This product provides existing RapidHIT ID customers a path to upgrade to the Applied Biosystems-branded RapidLINK Software v1.0.

Features of the Applied Biosystems RapidLINK Software v1.0 include:

  • Securely manage one or more Applied Biosystems RapidHIT ID Systems from a central location
  • Review DNA profile results in real time prior to upload
  • Enable or disable users in real time
  • Collect and review metadata using built-in reporting tools
  • Expert system-capable
  • Sample/data accessioning workflow-capable

This product includes:

  • Enveloped letter with upgrade/installation details

Specifications

Software Type Upgrade
Quantity 1 upgrade
Product Line RapidLINK
Shipping Condition Room Temperature
Content And Storage 1 document letter