missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Applied Biosystems™ RapidLINK™ Software v1.0 Staff Elimination Database Application

Catalog No. A41816
Change view
Click to view available options
Quantity:
1 annual license
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A41816 1 annual license
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A41816 Supplier Applied Biosystems™ Supplier No. A41816

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems RapidLINK Staff Elimination Database Application enables low-effort monitoring of possible contamination from laboratory and agency staff members, manufacturing employees, and other personnel who may come in contact with DNA during the process of generating a DNA profile.

The Applied Biosystems RapidLINK Staff Elimination Database Application enables low-effort monitoring of possible contamination from laboratory and agency staff members, manufacturing employees, and other personnel who may come in contact with DNA during the process of generating a DNA profile.

The matching function between profiles in the RapidLINK database and the staff database runs in the background at all times, alerting the RapidLINK Software user only when there is a possible contamination event. The RapidLINK Staff Elimination Database Application automatically generates a printable summary report identifying the profile for further review and resolution. For security of the elimination database, only RapidLINK administrators with the proper access levels can upload employee profiles or remove inactive ones to keep the elimination database current. For use only with Applied Biosystems RapidLink Software v1.0.

Features of the RapidLINK Staff Elimination Database Application include:

  • Identification of staff-related contamination
  • Meets a lab quality requirement
  • Reduced workload for lab management
  • Background application is always 'on'
  • Automatically generated report
  • Administrator-defined database

Specifications

Software Type License
Quantity 1 annual license
Product Line RapidLINK
For Use With (Equipment) Applied Biosystems RapidLINK Software V1.0
Product Type Staff Elimination Database Application Software
Shipping Condition Room Temperature
Content And Storage 1 document letter