missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Applied Biosystems™ RapidLINK™ Software v1.0 Match Application

Catalog No. A41818
Change view
Click to view available options
Quantity:
1 annual license
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A41818 1 annual license
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A41818 Supplier Applied Biosystems™ Supplier No. A41818

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems RapidLINK Match Application is an easy-to-use application software that provides automatic matching of DNA profiles in the RapidLINK database.

The Applied Biosystems RapidLINK Match Application is an easy-to-use application software that provides automatic matching of DNA profiles in the RapidLINK database. The application compares one reviewed profile to all other available profiles in a passive search, so that user-defined parameters are automatically applied to all match queries. For DNA profiles that may have a match, the RapidLINK Match Application generates a printable report that summarizes the likelihood of the overall profile match and details the probability of each allelic match. For use only with Applied Biosystems RapidLink Software v1.0.

Features of the RapidLINK Match Application include:

  • Powerful search tool
  • Flexible with regard to size of pool
  • Reduced workload for lab personnel
  • Passive search is always 'on'
  • User-defined search parameters
  • Automatically generated match report

Specifications

Software Type License
Quantity 1 annual license
Product Line RapidLINK
For Use With (Equipment) Applied Biosystems RapidLINK Software V1.0
Product Type Match Application Software
Shipping Condition Room Temperature
Content And Storage 1 document letter