missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ RapidLINK™ Software v1.0 License
$20,150.00 - $60,700.00
Specifications
| Product Line | RapidLINK |
|---|---|
| Software Type | License |
| Content And Storage | 1 document letter |
| Shipping Condition | Room Temperature |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
A41813
|
N/A | 1 user license |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
A41814
|
N/A | 3 user licenses |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
A41815
|
N/A | 10 user licenses |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Specifications
| RapidLINK | |
| License | |
| 1 document letter | |
| Room Temperature |