missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Applied Biosystems™ RapidLINK™ Software v1.0 Kinship Application
Description
The Applied Biosystems RapidLINK Kinship Application is an easy-to-use application software that confirms the relationship between two profiles on a three-generation family pedigree. The application generates a printable summary report of the likelihood ratio of the overall relationship and of the allelic relationship at each locus. For use only with Applied Biosystems RapidLink Software v1.0.
Features of the RapidLINK Kinship Application include:
- Confirmation of specific family relationship between two DNA profiles
- Flexible with regard to size of pool
- Reduced workload for lab personnel
- Calculates likelihood ratios of two DNA profiles on a family pedigree
- User-defined search stringency
- Automatically generated report
Specifications
Specifications
| Software Type | License |
| Quantity | 1 annual license |
| Product Line | RapidLINK |
| For Use With (Equipment) | Applied Biosystems RapidLINK Software V1.0 |
| Product Type | Kinship Application Software |
| Shipping Condition | Room Temperature |
| Content And Storage | 1 document letter |