missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Applied Biosystems™ RapidLINK™ Software v1.0 Familial Application

Catalog No. A41819
Change view
Click to view available options
Quantity:
1 annual license
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A41819 1 annual license
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A41819 Supplier Applied Biosystems™ Supplier No. A41819

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems RapidLINK Familial Application is an easy-to-use application software that generates a list of DNA profiles in the RapidLINK database that may be familiarly related to each other.

The Applied Biosystems RapidLINK Familial Application is an easy-to-use application software that generates a list of DNA profiles in the RapidLINK database that may be familiarly related to each other. The application generates a printable, summary report of profiles that may have a parent-child or sibling-sibling relationship to the profile being queried, and details the likelihood ratio of the overall relationship and the likelihood ratio of the allelic relationship at each locus. For use only with Applied Biosystems RapidLink Software v1.0.

Features of the RapidLINK Familial Application include:

  • Identification of familiarly related DNA profiles
  • Flexible with regard to size of pool
  • Reduced workload for lab personnel
  • Calculates likelihood ratios of parent-child and sibling-sibling relationships
  • User-defined search stringency
  • Automatically generated match report

Specifications

Software Type License
Quantity 1 annual license
Product Line RapidLINK
For Use With (Equipment) Applied Biosystems RapidLINK Software V1.0
Product Type Familial Application Software
Shipping Condition Room Temperature
Content And Storage 1 document letter