missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Random Hexamers (50 μM) DFS Item

Random Hexamers are short oligodeoxyribonucleotides of random sequence [d(N)6] that anneal to random complementary sites on a target DNA or RNA to serve as primers for DNA synthesis by a DNA polymerase or reverse transcriptase.

Supplier:  Applied Biosystems™ N8080127

Catalog No. N8080127


Only null left

Includes: Contains Random Hexamers (100 μL) at a concentration of 50 μM. Sufficient for 100 reverse transcriptions of 20 μL each.

Description

Description

Random Hexamers are short oligodeoxyribonucleotides of random sequence [d(N)6] that anneal to random complementary sites on a target DNA or RNA to serve as primers for DNA synthesis by a DNA polymerase or reverse transcriptase.

For this product five nmoles of primer are supplied at a concentration of 50 μM.

Applications
Most commonly used for complementary DNA (cDNA) synthesis priming, these random hexamers may be used for a variety of applications including:
• Two-step RT-qPCR
• cDNA Synthesis
• RT-PCR

Order Info

Shipping Conditions: Dry Ice

Specifications

Specifications

Primer
Dry Ice
Random Hexamer
DNA Synthesis
• Random Hexamers (100 μL at 50 μM), sufficient for 100 reverse transcriptions of 20 μL each

Store at –15 to –25°C.
50 μM
5 nmol, 100 μL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.