missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PCR Tubes & Caps, RNase-free, 0.2 mL (8-strip format)

Catalog No. am12230
Encompass
Change view
Click to view available options
Quantity:
125 Strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM12230 125 Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM12230 Supplier Applied Biosystems™ Supplier No. AM12230
Only null left

These 0.2mL PCR tubes with caps are provided in an 8-strip format and guaranteed RNase- and DNase-free.

These 0.2mL PCR tubes with caps are provided in an 8-strip format and guaranteed RNase- and DNase-free. Tubes and caps design allows caps to seal securely with minimal pressure. Polypropylene tubes are autoclave-safe.

Specifications

Color Clear
Volume (Metric) 0.2 mL
Quantity 125 Strips
For Use With (Equipment) Thermal Cyclers
Material Polypropylene
Content And Storage Store at room temperature.
Purity or Quality Grade RNase-Free, DNase-Free
Reaction Speed Standard
Shipping Condition Room Temperature
Closure Type 8-Strip Cap
Autoclavable Autoclavable
Closure Included Yes
Format Tube
Product Type PCR Tube and Cap
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.