missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Oligo d(T)16 (50 μM) DFS Item

Invitrogen Oligo(dT)16 Primer is a homo-oligomeric deoxyribonucleotide poly(dT) that is non-phosphorylated and chemically synthesized.

Supplier:  Applied Biosystems™ N8080128

Catalog No. N8080128

Only null left

Includes: Oligo(dT)16 primer is supplied in 100 μL of 10 mM TE, pH 7.0, at a concentration of 50 μM.

Description

Description

Invitrogen Oligo(dT)16 Primer is a homo-oligomeric deoxyribonucleotide poly(dT) that is non-phosphorylated and chemically synthesized. It is used for priming and reverse transcription of polyadenylated poly(A)+ mRNA.

Five nmoles of Oligo (dT)16 Primer is supplied in 100 μL of 10 mM TE, pH 7.0, at a concentration of 50 μM.

Order Info

Shipping Conditions: Dry Ice

Specifications

Specifications

Primer
Reverse Phase HPLC
50 μM
5 nmol, 100 μL
Liquid
• Oligo(dT)16 Primer (100 μL at 50 μM)

Store at –10 to –30°C.
Aliquot to avoid repeated freezing and thawing.
Dry Ice
Oligo dT
Reverse Transcription
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.