missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ Optical 384-Well Reaction Plate

Catalog No. 4343370
Encompass
Change view
Click to view available options
Quantity:
1000 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4343370 1000 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4343370 Supplier Applied Biosystems™ Supplier No. 4343370

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimized to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification

Constructed from a single rigid piece of polypropylene in a 384-well format, the plate is engineered to work with Applied Biosystems Real-Time PCR systems and GeneAmp™ PCR systems.

  • Frosted to minimize interfering fluorescence from wells of cycling block
  • Screened to eliminate autofluorescent plates
  • Design provides barrier to ambient air to help ensure well-to-well temperature uniformity

  • Platform: 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7900HT System, GeneAmp™ 9700, QuantStudio™, QuantStudio Dx, Veriti™ Dx Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA 7 System

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

Specifications

Barcode No Barcode
Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 1000 plates.

May be stored at room temperature. Under proper storage conditions, plates may be stored indefinitely.
Packaging 1,000 Plates
Product Line MicroAmp
Color Optical
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Skirted
Material Polypropylene
No. of Wells 384
Quantity 1000 plates
Reaction Speed Standard
Shipping Condition Room Temperature
Show More Show Less
Are the MicroAmp Optical 96-Well Reaction Plates and MicroAmp Optical 384 -Well Reaction Plates autoclavable?

Our microplates are certified to be DNA, RNase and PCR inhibitor free as long as they are kept in the original packaging bag. They are not meant to be autoclaved.