missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL

Catalog No. 4366932
Encompass
Change view
Click to view available options
Quantity:
20 Plates
200 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4366932 200 Plates
4346906 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4366932 Supplier Applied Biosystems™ Supplier No. 4366932

Reduces PCR reaction time from 2 hours to as little as 25 minutes

  • Integral component of Applied Biosystems Fast PCR System
  • Maximum thermal conductivity for precise thermal cycling
  • Reproducible, specific, and sensitive PCR results in just 25 minutes
  • Validated with other system components for consistent results
  • Each reaction plate includes unique serialized, eight character number label (barcode) that is user-readable and machine-readable to prevent tracking errors
  • Increased thermal contact for faster, more uniform heating
  • Advanced design dramatically increases its thermal contact with Applied Biosystems Veriti™ 96-Well Fast Thermal Cycler
  • Consistent well-to-well thickness helps all samples cycle through PCR process uniformly

  • Platform: 310 Genetic Analyzer, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7500 Fast Dx System, 7500 Fast System, 7900HT Fast System, StepOnePlus™, Veriti™ Dx Fast Thermal Cycler, Veriti Fast Thermal Cycler, ViiA™ 7 Dx Fast System, ViiA 7 Fast System

Applications: Fast Real-Time PCR, PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Ambient

Specifications

Barcode Includes Barcode
Certifications/Compliance Certified DNA/RNase Free
Content And Storage Contains 200 plates (equivalent to 10 × catalog # 4346906).

Store at room temperature. Under proper storage conditions, plates may be stored indefinitely.
Packaging 200 Plates
Product Line MicroAmp
Color Optical
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Semi
Material Polypropylene
Volume (Metric) Well 0.2 mL
No. of Wells 96
Volume (Metric) Working 0.1 mL
Quantity 200 Plates
Reaction Speed Fast
Show More Show Less
Are the MicroAmp Optical 96-Well Reaction Plates and MicroAmp Optical 384 -Well Reaction Plates autoclavable?

Our microplates are certified to be DNA, RNase and PCR inhibitor free as long as they are kept in the original packaging bag. They are not meant to be autoclaved.

Can I place an Optical Fast 96-Well plate onto a standard Applied Biosystems 7500 or 7300 Real-Time PCR System?

No. The fast plate will not fit correctly into the standard block and assay performance will be significantly compromised.

Can I use MicroAmp Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL (Cat. No. 4346906, 4366932) with the QuantStudio 7 Flex Real-Time PCR System in the Standard run mode?

Yes. MicroAmp Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL (Cat. No. 4346906, 4366932) can be used with either Fast or Standard run modes. However, the “Fast” plates can only be run on the 96-well Fast block, which is for smaller volumes rather than a standard block (0.2 mL). The Fast or Standard mode for the run is determined by the polymerase in the master mix used in the experiment. Please refer to the user guide for the specific master mix to determine which mode to use.