missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ Centrifuge Adapter

Catalog No. N8013822
Encompass
Change view
Click to view available options
Quantity:
1 Adaptor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N8013822 1 Adaptor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N8013822 Supplier Applied Biosystems™ Supplier No. N8013822

Includes

1 MicroAmp Centrifuge Adaptor

Designed to fit snugly into standard microcentrifuges for high-speed centrifugation of MicroAmp reaction tubes

Accommodates a single MicroAmp reaction tube, fits a standard microcentrifuge rotor, and can be used at up to 15,000 x g.

  • Accommodates single MicroAmp reaction tube for convenience
  • Constructed to fit standard microcentrifuge rotor, offering flexibility

See user's manual or package insert for limited label license and trademark information.

PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store at Room Temperature
For Use With (Equipment) 2720 Thermal Cycler, GeneAmp™ 9700, ProFlex™ PCR System, SimpliAmp™ Thermal Cycler, Veriti™ Dx Thermal Cycler, Veriti™ Thermal Cycler
Quantity 1 Adaptor

For Research Use Only. Not for use in diagnostic procedures.