missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MicroAmp™ 96-Well Tray
Specifically designed to snugly hold up to 96 MicroAmpReaction tubes with caps
Supplier: Applied Biosystems™ N8010541
Comprend: 10 MicroAmp 96-Well Bases.
Description
The MicroAmp 96-well tray is required for more efficient sample processing on Applied Biosystems96-well thermal cyclers.
- Engineered for efficient sample processing
- Designed specifically for MicroAmp reaction tubes with Caps
- Manufactured to high specification, ensuring quality product
- Allows to organize tubes while adding sample and mix
- Reduces spillage and tube mix-ups by securing and ordering your tubes on lab bench
- When samples are ready, tray of tubes are directly on block of thermal cycler
- Eliminates hassle of transferring tubes
- Provides thermal contact necessary for thermal cycling
See user's manual or package insert for limited label license and trademark information.
PCR, PCR and Real-Time PCR, Real Time PCR (qPCR)
Order Info
Shipping Condition: Room temperature
Spécifications
| Certified DNA/RNase Free | |
| MicroAmp | |
| Tray | |
| PCR | |
| 10 Trays |
| Contains 10 MicroAmp™ 96-Well Trays. Store at room temperature. | |
| Natural | |
| GeneAmp 9700, 2720 Thermal Cycler | |
| 96-Well Plates | |
| Room Temperature |