missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 8-Cap Strip, clear

Catalog No. N8011535
Encompass
Change view
Click to view available options
Quantity:
1500 Strips (5 pk./300)
300 Strips
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N8011535 1500 Strips (5 pk./300)
N8010535 300 Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N8011535 Supplier Applied Biosystems™ Supplier No. N8011535

Designed to fit on MicroAmp reaction tubes, tube strips, and 96-well plates

MicroAmp™ Cap Strips have a domed profile and are suitable for end-point PCR. 8-cap strips available in clear or an assorted set of red, orange, blue, and green.

  • Will not interfere with sample reading
  • Provides tight seal to minimize evaporation
  • Easy to apply and remove
Minimizes Evaporation
  • Use MicroAmp Caps Strips to seal tubes and reduce the possibility of contamination in sample wells and help ensure reliable results
  • Easy to Use
    • Use MicroAmp Cap Installing Tool (sold separately) to push caps down and form tight seal
    • When ready, just peel away caps using tabs at end
    Colored for Easy Organization
    • Available in different colors to help track tubes during assay preparation
    • For real-time PCR applications,use MicroAmp Optical 8-Cap Strips (Mfr. No. 4323032) with flat-top profile

    PCR, PCR & Real-Time PCR

    Specifications

    Certifications/Compliance Certified DNA/Rnase Free
    Color Natural
    Quantity 1500 Strips (5 pk./300)
    For Use With (Equipment) Thermal Cyclers
    Product Line MicroAmp™ 
    Material Polypropylene
    Content And Storage Includes: 5 packs of N8010535 (Qty 300 per pack)

    Strip of 8 attached caps for use with 0.2 mL individual tubes, strip tubes (0.1 or 0.2 mL), or 48- or 96-well plates. Natural (clear) color.
    Closure Type Domed Cap
    No. of Wells 96
    Format 1,500 Strip Caps
    Product Type 8-Strip Cap
    Show More Show Less
    Can I use caps on the 96-well plates on the QuantStudio 12K Flex system?

    Yes. We recommend that you use Optical Flat caps, such as Cat. No. N8010535. Use of domed caps may damage the heated cover.