missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 48-Well Optical Adhesive Film

Reduces the chance of well-to-well contamination and sample evaporation when applied to a 48-well microplate

Supplier:  Applied Biosystems™ 4375323

Encompass

 View more versions of this product

Catalog No. 4375323


Only null left

Includes: 100 or 25 MicroAmp 48-Well Optical Adhesive Films

Description

Description

  • Pressure-sensitive adhesive provides tight seal over every well
  • Will not interfere with sample reading
  • Easy to apply and remove, will not stick to gloves
  • Prevent Sample Loss
  • Optically clear adhesive film prevents sample loss by securely sealing into wells of microplates and reducing leaks, evaporation, and contamination between sample wells to help ensure consistent real-time PCR data
  • Easy to Use: simply peel backing and apply firmly across top of microplate
  • Once sealed, microplates can be safely stacked without worrying about contamination
  • Designed to fit MicroAmp Fast Optical 48-Well Plates
Note: See user's manual or package insert for limited label license and trademark information.

For Research Use Only. Not for use in diagnostic procedures.

Applications:

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

Specifications

Chemical Identifiers

Optical
48
Room Temperature
MicroAmp
48-well Plate
Polyester
StepOne™,
Film, Seal, or Applicator
Pressure Sensitive

Specifications

Optical
48
Contains 100 MicroAmp™ 48-Well Optical Adhesive Films. Store at room temperature.
Room Temperature
MicroAmp
48-well Plate
Polyester
100 films
StepOne™,
Film, Seal, or Applicator
Pressure Sensitive
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions