missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 12-Cap Strip DFS Item

Designed to fit on MicroAmp reaction tubes, tube strips, and 96-well plates

$191.00 - $764.00

Specifications

Certifications/Compliance Certified DNA/Rnase Free
Color Assorted
For Use With (Equipment) Thermal Cyclers
Product Line MicroAmp™
Material Polypropylene
View More Specs

Products 2
Catalog Number Mfr. No. Color Quantity For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Color Quantity For Use With (Equipment) Price Quantity  
N8010534 DFS Item
View Documents
Applied Biosystems™
N8010534
Assorted 200 Strips Thermal Cyclers
Each for $191.00
Only null left
 
N8011534 DFS Item
View Documents
Applied Biosystems™
N8011534
Assorted 1000 Strips (5 pk./200) Thermal Cyclers
Each for $764.00
Only null left
 
Description

Description

MicroAmp™ Cap Strips have a domed profile and are suitable for end-point PCR. 12-cap strips available in clear or an assorted set of red, orange, blue, and green.

  • Will not interfere with sample reading
  • Provides tight seal to minimize evaporation
  • Easy to apply and remove
Minimizes Evaporation
  • Use MicroAmp Caps Strips to seal tubes and reduce possibility of contamination in sample wells and help ensure reliable results
  • Easy to Use
  • Use MicroAmp Cap Installing Tool (sold separately) to push caps down and form tight seal
  • When ready, just peel away caps using tabs at end
  • Colored for Easy Organization
    • Available in different colors to help track tubes during assay preparation

    See user's manual or package insert for limited label license and trademark information.

    PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

    Specifications

    Specifications

    Certified DNA/Rnase Free
    Thermal Cyclers
    Polypropylene
    96
    Assorted
    MicroAmp™
    Domed Cap
    12-Strip Cap
    Videos
    SDS
    missing translation for 'documents'

    missing translation for 'documents'