missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 12-Cap Strip

Catalog No. N8011534
Encompass
Change view
Click to view available options
Quantity:
1000 Strips (5 pk./200)
200 Strips
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N8011534 1000 Strips (5 pk./200)
N8010534 200 Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N8011534 Supplier Applied Biosystems™ Supplier No. N8011534

Designed to fit on MicroAmp reaction tubes, tube strips, and 96-well plates.

MicroAmp™ Cap Strips are designed to fit on MicroAmp™ reaction tubes, tube strips, and 96-well plates. They are available in 8-cap and 12-cap strips in clear or an assorted set of red, orange, blue, and green. They have a domed profile and are suitable for end-point PCR. Attach caps using the MicroAmp™ Cap Installing Tool.

Will not interfere with sample reading.

  • Provides tight seal to minimize evaporation
  • Easy to apply and remove

Minimizes Evaporation

Use MicroAmp™ Caps Strips to seal the tubes containing your precious samples. This will reduce the possibility of contamination among sample wells and help ensure reliable results.

Easy to Use

Use the MicroAmp™ Cap Installing Tool to push the caps down and form a tight seal. When ready, just peel away the caps using the tabs at the end.

Colored for Easy Organization

MicroAmp™ Caps Strips are available in different colors to help you keep track of your tubes during assay preparation.

Note: See user's manual or package insert for limited label license and trademark information.

Specifications

Certifications/Compliance Certified DNA/Rnase Free
Color Assorted
Quantity 1000 Strips (5 pk./200)
For Use With (Equipment) Thermal Cyclers
Product Line MicroAmp™
Material Polypropylene
Content And Storage Contains 1,000 MicroAmp™ 12-Cap Strips (5 packs of SKU# N8010534, Qty 200 per pack). Store at room temperature.
Closure Type Domed Cap
No. of Wells 96
Format 1,000 Strip Caps
Product Type 12-cap Strip
Show More Show Less