missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 12-Cap Strip, assorted colors

Catalog No. N8010834
Encompass
Change view
Click to view available options
Quantity:
200 Strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N8010834 200 Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N8010834 Supplier Applied Biosystems™ Supplier No. N8010834

Designed to fit on MicroAmp reaction tubes, tube strips, and 96-well plates

MicroAmp™ Cap Strips have a domed profile and are suitable for end-point PCR. 12-cap strips available in clear or an assorted set of red, orange, blue, and green.

 

  • Will not interfere with sample reading
  • Provides tight seal to minimize evaporation
  • Easy to apply and remove
Minimizes Evaporation
  • Use MicroAmp Caps Strips to seal tubes and reduce possibility of contamination in sample wells and help ensure reliable results
  • Easy to Use
  • Use MicroAmp Cap Installing Tool (sold separately) to push caps down and form tight seal
  • When ready, just peel away caps using tabs at end
  • Colored for Easy Organization
  • Available in different colors to help track tubes during assay preparation

 

See user's manual or package insert for limited label license and trademark information.

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

Specifications

Certifications/Compliance Certified DNA/Rnase Free
Color Assorted
Volume (Metric) 0.2 mL
Quantity 200 Strips
For Use With (Equipment) Thermal Cyclers
Product Line MicroAmp™ 
Material Polypropylene
Content And Storage Contains 200 MicroAmp™ 12-Cap Strips, Assorted Colors. Store at room temperature.
Shipping Condition Room Temperature
Closure Type Domed Cap
No. of Wells 96
Format 200 Strip Caps
Product Type 12-Strip Cap
Show More Show Less