missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ JUN™ Dye Spectral Calibration Plate for Multiplex qPCR, 384-well DFS Item

Used for JUN™ dye spectral calibration of your ViiA™ 7, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, and QuantStudio™ 12K Flex real-time PCR systems with a 384-well block.

Supplier:  Applied Biosystems™ A24733

Catalog No. A24733


Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Only null left

Description

Description

Calibration and verification should be run at least every six months. Please see your instrument maintenance guide for full instructions on use. The dye formulations will improve results with multiplexing by more accurately representing fluorescent spectra found in your real-time PCR experiments.

Calibration and verification should be run at least every six months. See instrument maintenance guide for full instructions. The dye formulation will improve results with multiplexing by more accurately representing fluorescent spectra found in real-time PCR experiments.

Order Info

Store in freezer (-5° to -30°C)

Specifications

Specifications

Store in Freezer (-5°C to -30°C).
QuantStudio™, ViiA™ 7 System
Real Time PCR (qPCR)
Spectral Calibration
JUN™, JUN™
Dye Spectral Calibration Plate
1 plate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.