missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ Thin-Walled Reaction Tube, with domed cap, 0.5 mL, autoclaved

Catalog No. N8010611
Encompass
Change view
Click to view available options
Quantity:
1000 Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N8010611 1000 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N8010611 Supplier Applied Biosystems™ Supplier No. N8010611

Includes

1000 GeneAmp thin-walled reaction tubes with domed cap, 0.5mL, autoclaved

Autoclaved to provide clean, controlled start

Thin-walled tubes facilitate rapid heat transfer to and from samples to achieve faster cycle times and increased specificity for precise PCR in 0.5mL PCR Systems.

  • Thin plastic enables fast heating and cooling for accurate results
  • Flat, frosted caps for easy marking
  • Captive lid with positive-click closure assures proper seating
  • Polished surface and conical bottom allow maximum sample recovery
  • Also available in economical bulk packs
  • Rapid Heat Transfer
  • Specially engineered polypropylene optimized for use with Veriti™ 60-Well Thermal Cycler and Dual 96-Well GeneAmp PCR System 9700. Tubes undergo stringent quality testing to ensure reproducible results
  • Easy to Use
  • Tubes feature flat, frosted cap, tight seal, and clear conical bottom
  • Recommended maximum reaction volume is 100μL

See user's manual or package insert for limited label license and trademark information.

PCR, PCR and Real-Time PCR

Specifications

Certifications/Compliance Certified DNA/Rnase Free
Color Natural
Volume (Metric) Working 0.5 mL
Volume (Metric) 0.6 mL
Quantity 1000 Tubes
For Use With (Equipment) Thermal Cyclers
Product Line GeneAmp
Sterility Sterile
Material Polypropylene
Content And Storage Contains 1,000 GeneAmp™ Thin-Walled Reaction Tubes with Domed Cap, 0.5 ml, Autoclaved. Store at room temperature.
Reaction Speed Fast
Shipping Condition Room Temperature
Closure Type Domed Cap
No. of Wells 96
Autoclavable Autoclavable
Format 1,000 Tubes
Product Type Reaction Tube
Show More Show Less