Learn More
Applied Biosystems™ GeneAmp™ dNTP Blend with dUTP (12.5 mM)
Description
GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. GeneAmp dNTPs with dUTP can be used to generate dU-containing PCR products in a carry-over prevention strategy in conjunction with AmpErase Uracil N-glycosylase (UNG).
Highlights
- High purity dNTPs are ideal for use in PCR or other applications
- dNTPs with dUTP are optimized for use in carry-over prevention
- Guaranteed PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
- Premixed dNTP blends are available in various concentrations, offering ease of use
Notes
GeneAmp dNTP Blend, 12.5 mM with dUTP contains 2.5 mM each of dATP, dCTP, dGTP and 5.0 mM of dUTP
Order Info
Shipping Condtion: Dry Ice
The PCR process is covered by patents owned by Roche Molecular Systems, Inc. and F. Hoffmann-La Roche Ltd.
Specifications
Specifications
| Concentration | 12.5 mM |
| Content And Storage | GeneAmp dNTP Blend with dUTP, 12.5 mM (1 mL) Store at –20°C. |
| Form | Liquid |
| Product Line | GeneAmp |
| Quantity | 1 mL |
| Shipping Condition | Dry Ice |
| For Use With (Application) | Standard PCR |
Frequently Asked Questions (FAQs)
Thymine is normally found as a deoxyribose. Thus, TTP and dTTP are essentially the same thing. Both types of mixes can be used in PCR reactions
You will need to confirm with the manufacturer of the restriction enzyme you are using to find out whether or not it can recognize dUTP-containing DNA.
For Research Use Only. Not for use in diagnostic procedures.