missing translation for 'onlineSavingsMsg'
Learn More

Applicant Fingerprint Cards

Catalog No. S69015
Change view
Click to view available options
Color:
Brilliant White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S69015 Brilliant White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S69015 Supplier Customer Channels Group (Ccg) Supplier No. 5993

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provide a brilliant white finish and fast ink absorption.

  • 110 lb. Paper stock
  • Standard format
  • Pack of 100

Specifications

Color Brilliant White
For Use With (Application) Provide a brilliant white finish and fast ink absorption
Product Type Applicant Fingerprint Cards