missing translation for 'onlineSavingsMsg'
Learn More

Promega ApoTox-Glo™ Triplex Assay System

Catalog No. PRG6321
Encompass
Change view
Click to view available options
Quantity:
10 mL
5 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG6321 5 x 10 mL
PRG6320 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRG6321 Supplier Promega Supplier No. G6321

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The ApoTox-Glo™ Triplex Assay combines three assay chemistries to easily assess viability, cytotoxicity and apoptosis events in the same cell-based assay well.

  • Assess Viability, Cytotoxicity and Apoptosis in One Assay
  • Easy to implement with an add-mix-measure protocol
  • Normalize data with a built-in control
  • Flexible and easily automated

Specifications

Quantity 5 x 10 mL
Content And Storage 2 x 10 mL Assay Buffer, 50 μL GF-AFC substrate, 50 μL bis-AAF-R110 substrate, 5 x 10 mL Caspase-Glo™ 3/7 Buffer, 5 x 1 bottle Caspase-Glo 3/7 substrate. Store at -30°C to -10°C

  • (a)U.S. Pat. Nos. 7,416,854, 7,553,632 and other patents pending.
  • (b)U.S. Pat. Nos. 6,602,677 and 7,241,584, Australian Pat. Nos. 754312 and 785294 and other patents and patents pending.
  • (c)The method of recombinant expression of Coleoptera luciferase is covered by U.S. Pat. Nos. 5,583,024, 5,674,713 and 5,700,673.