missing translation for 'onlineSavingsMsg'
Learn More

M&I Materials Apiezon™ Wax

Hard permanent vacuum sealing/mounting wax and etch resist

Supplier:  M&I Materials FAPWW00500

Catalog No. 14-638-25C



Only null left

Description

Description

  • Vapor pressure less than 8 × 10-7 Pa (4.5 × 10-9 torr) at 20°C
  • Softening point, 80° to 90°C (176° to 194°F)
  • Resistant to etching fluids

Order Info

Supplied in packs of 25 sticks; total weight 1 lb. (500g).

Specifications

Specifications

Wax Stick
Wax
500 Pk.
80°C to 90°C
500 g
Hard permanent vacuum sealing/mounting wax and etch resist
4.5 x 109 torr
176°F to 194°F
1 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions