Learn More
Electron Microscopy Sciences Apiezon Vacuum Sealing Wax W Hard Vacuum Sealing 500 GR
Catalog No. 501902647
Encompass
Easy to use Apiezon waxes can be used as permanent or semi-permanent vacuum sealants or mounting media. The waxes have a high purity combined with powerful gettering and clean meltdown properties. An excellent waterproof media that is easy to use and to remove. 10 year shelf life. Wax w is a hard vacuum sealing and mounting wax that is etch resist.
Use: Hard Vacuum Sealing / Mounting Wax, Etch Resist
Catalog No. 501902647 Supplier Electron Microscopy Sciences Supplier No. 60710
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Easy to use Apiezon waxes can be used as permanent or semi-permanent vacuum sealants or mounting media. The waxes have a high purity combined with powerful gettering and clean meltdown properties. An excellent waterproof media that is easy to use and to remove. 10 year shelf life. Wax w is a hard vacuum sealing and mounting wax that is etch resist.
Use: Hard Vacuum Sealing / Mounting Wax, Etch Resist