missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ AP800 ESD Nitrile-Coated Gloves

Catalog No. 18139747
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18139747 Small
18139746 Medium
18139745 Large
18139748 X-Large
18139749 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18139747 Supplier SHOWA™ Supplier No. AP800L08
Only null left

Electrostatic dissipative 13-gauge micro fiber polyester, embossed microporous foam nitrile coating glove

The new SHOWA AP800 has a 13-gauge seamless knit with a Microfiber polyester static dissipative yarn and an embossed microporous foam nitrile palm dip. It is free of natural rubber latex and is touchscreen compatible, silicone-free and fingerprint-free. Certified under the standard EN 16350, the AP800 by SHOWA meets the requirements for electrostatic properties (ESD). This certification ensures the glove’s safe usage in ATEX zones (working area with risk of explosion and/or high flammability).

Specifications

Accelerator Free Yes
Applications Industrial
Coating Material Nitrile Foam
Color Gray
Disposable Reusable
Exterior Finish No Texture
Hand Ambidextrous
Latex Free Yes
Material Polyester
Powdered Powder Free
Product Type ESD Gloves
Size Small
Sterility Non-sterile
Thickness (English) 13 gauge
Thickness (Metric) 0.7 mm
Show More Show Less