missing translation for 'onlineSavingsMsg'
Learn More

Antipyrine, 99%

Catalog No. AC104971000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC104971000 100 g
AC104975000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104971000 Supplier Thermo Scientific Chemicals Supplier No. 104971000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60-80-0 | C11H12N2O | 188.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60-80-0
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD00003146
InChI Key VEQOALNAAJBPNY-UHFFFAOYSA-N
Synonym antipyrine, phenazone, antipyrin, phenazon, analgesine, anodynine, anodynin, azophen, fenazone, 2,3-dimethyl-1-phenyl-5-pyrazolone
PubChem CID 2206
ChEBI CHEBI:31225
IUPAC Name 1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=CC(=O)N(N1C)C2=CC=CC=C2

Specifications

Melting Point 109°C to 113°C
Color White
Boiling Point 319°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Iodimetry)
Packaging Plastic bottle
Quantity 100 g
Beilstein 24,27
Merck Index 15,703
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and chloroform,slightly soluble in ether
Formula Weight 188.23
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Antipyrine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye prot
EINECSNumber 200-486-6
RTECSNumber CD2450000
TSCA TSCA

RUO – Research Use Only