missing translation for 'onlineSavingsMsg'
Learn More

Antimycin A

Catalog No. p-7058241
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63522LB0 5 mg
AAJ63522MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63522LB0 Supplier Thermo Scientific Chemicals Supplier No. J63522LB0

CAS: 1397-94-0 | C28H40N2O9 | 548.633 g/mol

Antimycin A is used to study the specific sites of reactive oxygen species production in mitochondria isolated from skeletal muscle of chronic obstructive pulmonary disease patients, and its relationship with local oxidative stress induced by exercise.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimycin A is used to study the specific sites of reactive oxygen species production in mitochondria isolated from skeletal muscle of chronic obstructive pulmonary disease patients, and its relationship with local oxidative stress induced by exercise.

Solubility
Soluble in 100% ethanol. Insoluble in water.

Notes
Isolated from Streptomyces sp.

Chemical Identifiers

CAS 1397-94-0
Molecular Formula C28H40N2O9
Molecular Weight (g/mol) 548.633
MDL Number MFCD01779723
InChI Key UIFFUZWRFRDZJC-RBVQMQRASA-N
PubChem CID 16218979
IUPAC Name [(2R,3S,6S,7R)-3-[(3-formamido-2-hydroxybenzoyl)amino]-8-hexyl-2,6-dimethyl-4,9-dioxo-1,5-dioxonan-7-yl] 3-methylbutanoate
SMILES CCCCCCC1C(C(OC(=O)C(C(OC1=O)C)NC(=O)C2=C(C(=CC=C2)NC=O)O)C)OC(=O)CC(C)C

Specifications

Type Antimycin A
Physical Form Powder
Melting Point 149°C to 150°C
Color White to Yellow
Quantity 5 mg
Sensitivity Light sensitive
UN Number UN3462
Merck Index 14,714
Solubility Information Soluble in 100% ethanol. Insoluble in water.
Formula Weight 548.63
Chemical Name or Material Antimycin A
Show More Show Less
Hazard Category H300+H310
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement P262-P264b-P270-P280-P301+P310-P302+P350-P310-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.
RTECSNumber CD0350000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only