missing translation for 'onlineSavingsMsg'
Learn More

Antimony(V)-chloride, 99%, anhydrous

CAS: 7647-18-9 | Cl5Sb | 299.02 g/mol

$113.30 - $370.87

Chemical Identifiers

CAS 7647-18-9
Molecular Formula Cl5Sb
Molecular Weight (g/mol) 299.02
MDL Number MFCD00011213
InChI Key VMPVEPPRYRXYNP-UHFFFAOYSA-I
Synonym antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
PubChem CID 24294
IUPAC Name pentachloro-$l^{5}-stibane
SMILES Cl[Sb](Cl)(Cl)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC201210500
SDS View Documents
Thermo Scientific Chemicals
201210500
50 g Glass bottle
Each for $113.30
Only null left
 
AC201212500
SDS View Documents
Thermo Scientific Chemicals
201212500
250 g Glass bottle
Each for $370.87
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Chemical Identifiers

7647-18-9
299.02
VMPVEPPRYRXYNP-UHFFFAOYSA-I
24294
Cl[Sb](Cl)(Cl)(Cl)Cl
Cl5Sb
MFCD00011213
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
pentachloro-$l^{5}-stibane

Specifications

92.0°C (30.0 mmHg)
7647-18-9
Liquid
98.5% min
SbCl5
01,42; 04,21; 05,18; 06,22; 07,12; 15,14; 17,17
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
VMPVEPPRYRXYNP-UHFFFAOYSA-I
pentachloro-$l^{5}-stibane
24294
99%
2.36
Antimony(V)-chloride, 99%
2.8°C
Colorless to Yellow
50 g
Cl5Sb
MFCD00011213
15, 686
Solubility in water: reacts with water forming hcl and antimonyoxide. Other solubilities: soluble in hcl, carbon disulfide, dichloromethane, soluble in an aqueous solution of tartaric acid,, ccl4 and chloroform
Cl[Sb](Cl)(Cl)(Cl)Cl
299.02
299.01
Glass bottle
2.3600g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 231-601-8

RTECSNumber : CC5075000

TSCA : TSCA

Recommended Storage : Product may develop some turbidity or precipitate

SDS
Documents

Documents

RUO – Research Use Only