missing translation for 'onlineSavingsMsg'
Learn More

Antimony(V) chloride, 99.997% (metals basis)

Catalog No. p-7049660
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1757014 25 g
AA1757022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1757014 Supplier Thermo Scientific Chemicals Supplier No. 01757014
Only null left

CAS: 7647-18-9 | Cl5Sb | 299.02 g/mol

Antimony(V) chloride is used as a catalyst and an analytical reagent for testing alkaloids and cesium. It is also used as a polymerization catalyst as well as involved in the chlorination of organic compounds. It acts as a Lewis acid and a strong oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7647-18-9
Molecular Formula Cl5Sb
Molecular Weight (g/mol) 299.02
MDL Number MFCD00011213
InChI Key VMPVEPPRYRXYNP-UHFFFAOYSA-I
Synonym antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
PubChem CID 24294
IUPAC Name antimony pentachloride
SMILES Cl[Sb](Cl)(Cl)(Cl)Cl

Specifications

Boiling Point 79°C (22mmHg)
Melting Point 2.8°C
Color Red-Yellow
Physical Form Liquid
Quantity 25 g
Linear Formula SbCl5
UN Number UN1730
Merck Index 14,695
Solubility Information Miscible with chloroform,hydrochloric acid,tartaric acid,carbon disulfide and carbon tetrachloride.
Formula Weight 299.02
Percent Purity 99.997%
Odor Pungent
Refractive Index 1.601
Sensitivity Moisture sensitive
Density 2.336 g/mL
Assay (metals basis)
Chemical Name or Material Antimony(V) chloride
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANTIMONY PENTACHLORIDE, LIQUID
EINECSNumber 231-601-8
RTECSNumber CC5075000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only