missing translation for 'onlineSavingsMsg'
Learn More

Antimony(V)-chloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

Catalog No. p-4228351
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434521000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC434521000 Supplier Thermo Scientific Chemicals Supplier No. 434521000
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Antimony(V)-chloride
Color Yellow
CAS 7647-18-9, 75-09-2
Physical Form Solution
Quantity 100 mL
Molecular Formula Cl5Sb
Linear Formula SbCl5
MDL Number MFCD00011213
Merck Index 15,686
Synonym antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
InChI Key VMPVEPPRYRXYNP-UHFFFAOYSA-I
SMILES Cl[Sb](Cl)(Cl)(Cl)Cl
IUPAC Name pentachloro-$l^{5}-stibane
Molecular Weight (g/mol) 299.02
PubChem CID 24294
Formula Weight 299.01
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.442
Density 1.4420g/mL
Show More Show Less

RUO – Research Use Only