missing translation for 'onlineSavingsMsg'
Learn More

Antimony Trichloride, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7165032
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608906 25 g Amber Glass Bottle
18608907 100 g Amber Glass Bottle
18608908 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18608906 Supplier Spectrum Chemical Mfg Cor Supplier No. A132025GM
Only null left

Chemical Identifiers

CAS 10025-91-9

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.