missing translation for 'onlineSavingsMsg'
Learn More

Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%

Catalog No. AA4061936
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4061936 500 g
AA4061914 25 g
AA4061922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4061936 Supplier Thermo Scientific Chemicals Supplier No. 04061936

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28300-74-5 | C8H4K2O12Sb2·3H2O

Antimony potassium tartrate trihydrate is used as a antiparasitic agent, which used in the treatment of schistosomiasis and leishmaniasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28300-74-5
Molecular Formula C8H4K2O12Sb2·3H2O
MDL Number MFCD00148863
Synonym Potassium antimonyl tartrate

Specifications

Physical Form Powder
Quantity 500 g
Assay Percent Range 99.0-103.0%
Solubility Information Soluble in water and glycerine. Insoluble in ethanol.
Formula Weight 667.87 (613.82 Anhydrous)
Grade ACS
Density 2.6
Chemical Name or Material Antimony potassium tartrate trihydrate
Hazard Category H302+H332-H335-H373
Hazard Statement Avoid release to the environment. Refer to special instructions/Safety data sheets
Precautionary Statement P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 229-436-1
RTECSNumber CC6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only