missing translation for 'onlineSavingsMsg'
Learn More

Antimony potassium tartrate hydrate, 98%

Catalog No. AAA1088936
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1088936 500 g
AAA1088922 100 g
AAA108890E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1088936 Supplier Thermo Scientific Chemicals Supplier No. A1088936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 331753-56-1 | C8H4K2O12Sb2 | 613.83 g/mol

Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Incompatible with mineral acids, strong bases, carbonates, lead, silver salts and strong oxidizing agents.

Chemical Identifiers

CAS 331753-56-1
Molecular Formula C8H4K2O12Sb2
Molecular Weight (g/mol) 613.83
MDL Number MFCD00148863
InChI Key GUJUCWZGYWASLH-UHFFFAOYNA-J
Synonym potassium antimony tartrate hydrate, potassium antimony iii tartrate hydrate, acmc-20ajv8, antimony 3+ potassium hydrate ditartrate
IUPAC Name dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate
SMILES [K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2

Specifications

Density 2.607
Assay Percent Range 98%
Quantity 500 g
UN Number UN1551
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 613.82 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Antimony potassium tartrate hydrate
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY POTASSIUM TARTRATE
EINECSNumber 608-190-2
RTECSNumber CC7350000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only