missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony pieces, 5mm (0.2in) & down, 99.9999% (metals basis)
CAS: 7440-36-0 | Sb | 121.76 g/mol
Supplier: Thermo Scientific Chemicals 01167618
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Silver to Gray | |
| 7440-36-0 | |
| 50 g | |
| 99.9999% (Metals basis) | |
| Sb | |
| powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german | |
| [Sb] | |
| 121.76 | |
| CHEBI:30304 |
| Antimony | |
| Silver Gray pieces | |
| Odorless | |
| Pieces | |
| MFCD00011211 MFCD00134030 | |
| WATWJIUSRGPENY-UHFFFAOYSA-N | |
| antimony | |
| 5354495 |
Chemical Identifiers
| 7440-36-0 | |
| 121.76 | |
| WATWJIUSRGPENY-UHFFFAOYSA-N | |
| 5354495 | |
| antimony |
| Sb | |
| MFCD00011211 MFCD00134030 | |
| powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german | |
| CHEBI:30304 | |
| [Sb] |
Safety and Handling
EINECSNumber : 231-146-5
RTECSNumber : CC4025000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only